Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 0.5 N

Ammonia solution 0.5 N

Plastic bottle, 2.500 mL
Catalog # KRAF03277.4000
Supplier:   Bernd Kraft
Ammonia solution 0.5 N
Catalog # KRAF03277.4000
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 0.5 N

Plastic bottle, 2.500 mL

CAS Number:  1336-21-6
Price
196,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2.500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.