Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 5% analytical reagent

Ammonia solution 5% analytical reagent

Plastic bottle, 5 L
Catalog # KRAF12389.5020
Irritant
Supplier:   Bernd Kraft
Ammonia solution 5% analytical reagent
Catalog # KRAF12389.5020
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 5% analytical reagent

Plastic bottle, 5 L

CAS Number:  1336-21-6
Price
113,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-647-6
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.