Ammonia solution 9,9 - 10,3% analytical reagent
Plastic bottle, 1 L
KRAF06212.3010
: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
- :1336-21-6
- :NH₃
- :MFCD00066650
- :35.05 g/mol
- :0.92 g/cm³ (20 °C)
- :Ambient
- :215-647-6
- :2672
- :8,III
- Pack type:Plastic bottle
- Pk:1 L