Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 32% analytical reagent

Ammonia solution 32% analytical reagent

Glass bottle, 1 L
Catalog # KRAF20068.3710
Corrosive
Irritant
Environment
Supplier:   Bernd Kraft
Ammonia solution 32% analytical reagent
Catalog # KRAF20068.3710
Supplier:  Bernd Kraft
CAS Number:  1336-21-6
Ammonia solution 32% analytical reagent

Glass bottle, 1 L

CAS Number:  1336-21-6
Price
52,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.