Buffer solution, colour coded, ready to use
7,0, Phosphate, Green, 20 °C, Plastic bottle, 5 L
KRAF03786.5400
- NIST traceable
- Colour coded with choice of colours for pH values 4,00 and 7,00
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:5 L
- Reference temperature:20 °C
- Colour:Green
- pH value:7,0



