Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methyl red

Methyl red

Plastic bottle for solids, 100 g
Catalog # KRAF17776.0600
Supplier:   Bernd Kraft
Methyl red
Catalog # KRAF17776.0600
Supplier:  Bernd Kraft
CAS Number:  493-52-7
Methyl red

Plastic bottle for solids, 100 g

CAS Number:  493-52-7
Price
165,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g

Specifications