Buffer solutions, ready to use
3,0, Citrate, Colourless, 20 °C, Plastic bottle, 10 L
Catalog # KRAF03082.6000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
- NIST traceable
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:10 L
- Reference temperature:20 °C
- Colour:Colourless
- pH value:3,0



