Order Entry
Belgium
Orders LinkContactUsLinkComponent
HiLoad Superdex columns, 75 PG

HiLoad Superdex columns, 75 PG

HiLoad™ 26/600 Superdex™ 75 PG, Glass, 34 µm, 26×600 mm, 320 ml, ≤ 13 ml, <2,6 ml/min, 10 - 50 cm/h
Catalog # 28-9893-34
Supplier:   Cytiva
HiLoad Superdex columns, 75 PG
Catalog # 28-9893-34
Supplier:  Cytiva
HiLoad Superdex columns, 75 PG
HiLoad Superdex columns, 75 PG

HiLoad™ 26/600 Superdex™ 75 PG, Glass, 34 µm, 26×600 mm, 320 ml, ≤ 13 ml, <2,6 ml/min, 10 - 50 cm/h

Price
4 400,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component

  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • 1/16" fittings for convenient connections to ÄKTA design systems

Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr < 10 000 (Superdex 30 pg), Mr 3 000 to 70 000 (Superdex 75 pg), and Mr 10 000 600 000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.

Delivery information: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.

Specifications
  • Flow rate:
    <2,6 ml/min
  • Particle Size:
    34 µm
  • Capacity:
    320 ml
  • Description:
    HiLoad™ 26/600 Superdex™ 75 PG
  • Seq:
    04
  • Column hardware:
    Glass
  • Bed dimensions:
    26×600 mm
  • Flow velocity:
    10 - 50 cm/h
  • Sample volume:
    ≤ 13 ml
  • Cat. No.:
    28-9893-34
  • Max. pressure:
    3 bar
  • Storage:
    4 to 30 °C, 20% Ethanol
  • pH range:
    3 - 12