ContactUsLinkComponent
Refrigerated bath/circulator, ltc4 kit. optima tc150 + r4 refrigeration unit, 20l capacity, inc. drain tap, insulated tubing and cli ps. temp range -30 to 100C. 1 * 1 items
Catalog # 461-0769
Supplier:  GRANT INSTRUMENTS
Refrigerated bath/circulator, ltc4 kit. optima tc150 + r4 refrigeration unit, 20l capacity, inc. drain tap, insulated tubing and cli ps. temp range -30 to 100C. 1 * 1 items
Catalog # 461-0769
Supplier:  GRANT INSTRUMENTS