ContactUsLinkComponent
Stirred bath/circulator, optima tc120 immersion thermostat + p12 plastic tank, 12l capacity. temp range amb.+5 to 99c. incl. integr ated pump. 1 * 1 items
Catalog # GRANTC120-P12EU
Supplier:  GRANT INSTRUMENTS
Stirred bath/circulator, optima tc120 immersion thermostat + p12 plastic tank, 12l capacity. temp range amb.+5 to 99c. incl. integr ated pump. 1 * 1 items
Catalog # GRANTC120-P12EU
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together