Agarose Basic
Glass bottle for solids, 1 kg
Catalog # A8963.1000
Supplier: PanReac AppliChem
Aller à :
Product Details & Documents
Specifications:
DNases/RNases/Proteases: not detectable
Ash: ≤ 0.45 %
Electroendoosmosis (EEO): 0.14 - 0.16
Gel strength (1 %): ≥ 1000 g/cm2
Gel strength (1.5 %): ≥ 2200 g/cm2
Gel point: 36 ± 1.5°C
Melting point: 88 ± 1.5°C
Clarity: max. 4 NTU
Sulfate: ≤ 0.15 %
- Numéro CAS:9012-36-6
- Numéro MDL:MFCD00081294
- Point de fusion:60…90 °C
- Température de stockage:Ambient
- EINECS:232-731-8
Spécifications
Spécifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



