Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agarose Basic

Agarose Basic

Glass bottle for solids, 250 g
Catalog # A8963.0250
Supplier:   PanReac AppliChem
Agarose Basic
Catalog # A8963.0250
Supplier:  PanReac AppliChem
CAS Number:  9012-36-6
Agarose Basic
Agarose Basic

Glass bottle for solids, 250 g

CAS Number:  9012-36-6
Price
205,00 €
Log in for availability
1 * 250 g
Product Details & Documents

Specifications:

DNases/RNases/Proteases: not detectable
Ash: ≤ 0.45 %
Electroendoosmosis (EEO): 0.14 - 0.16
Gel strength (1 %): ≥ 1000 g/cm2
Gel strength (1.5 %): ≥ 2200 g/cm2
Gel point: 36 ± 1.5°C
Melting point: 88 ± 1.5°C
Clarity: max. 4 NTU
Sulfate: ≤ 0.15 %

  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications