Order Entry
Belgium
Orders LinkContactUsLinkComponent
Micro extraction by packed sorbent (MEPS™)

Micro extraction by packed sorbent (MEPS™)

54,1 mm scale, 6,5 mm OD barrel, 250 µl, Agilent 7693A systems
Catalog # 549-0734
Micro extraction by packed sorbent (MEPS™)
Catalog # 549-0734
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)

54,1 mm scale, 6,5 mm OD barrel, 250 µl, Agilent 7693A systems

Price
199,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.

  • The BIN contains the SPE phase that is built into the syringe needle
  • The BIN is used with a 100 or 250 µl gas tight MEPS™ syringe that allows fluid handling at normal SPE pressures
  • Sample preparation for complex biological samples is readily adapted to MEPS™ and reduces the volume of sample and reagents required for extraction

Base material is silica with mean particle size of 45 µm and pore size of 60 Å.

Specifications
  • For:
    Agilent 7693A systems
  • Volume:
    250 µl
  • Description:
    54,1 mm scale, 6,5 mm OD barrel
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.