Accessories
Seal rubber on bottom inside door, G 788 (inc CD), PG 8535
MIEL1102422
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
- Description:Seal rubber on bottom inside door
- Seq:31
- For:G 788 (inc CD), PG 8535