ClipTip™ Racks for Tips
Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)
613-2221
Utilize Thermo Scientific™ ClipTip™ Pipette tip Racks, available in three sizes.
- Small rack: For use with ClipTip™ 20 and 50 Pipette tips
- Medium rack: For use with ClipTip™ 200 and 300 Pipette tips
- Large rack: For use with ClipTip™ 1000 Pipette tips
: Racks are supplied without tips and inserts.
- Description:Empty rack, medium, colour code yellow and orange (for ClipTip 200 and 300)
- Seq:03
- Cat. No.:613-2221
- Environmental certification:ISO 14001 (Certificate Registration No. EMS 623400)
- Environmentally Preferable:Low Manufacturing Impact
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.