Orders LinkContactUsLinkComponent
Reusable earplugs, E-A-R™ Express™
Reusable earplugs, E-A-R™ Express™
Polyurethane earplug, without cord, Yellow/blue, Universal
Catalog # 111-1865
Supplier:   3M
Reusable earplugs, E-A-R™ Express™
Catalog # 111-1865
Supplier:  3M
Reusable earplugs, E-A-R™ Express™
Reusable earplugs, E-A-R™ Express™
Polyurethane earplug, without cord, Yellow/blue, Universal
Price
105,00 €
Log in for availability
1 * 100 PAIRE
Product Details & Documents
E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can reduce noise levels by 28 dB, and have a hygienic insertion grip.
  • Easy to slide gently into ears with slow recovery foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
Protect hearing at work or during leisure activities with the E-A-R™ Express™ Reusable Earplugs. Designed to slide gently into the ear, the specially designed pod foam tip provides a comfortable and effective seal. The insertion grip eliminates the need to touch the foam, making them extremely hygienic. Get the best of both worlds with a patented slow recovery foam for a comfortable acoustic seal with quick and easy push-to-fit insertion.
Available in a recognisable yellow and blue for easy identification. Provides noise reduction to 28 dB SNR and fully tested and approved against EN 352-2. Model is compatible for use with the e 3M™ E-A-RFit™ Dual-Ear Validation System and available as a probed plug.
Certifications: EN 352-2
Attention: * One size fits all.
Specifications
  • Colour:
    Yellow/blue
  • Description:
    Polyurethane earplug, without cord
  • Seq:
    02
  • Size:
    Universal