Bottles, narrow neck, with thread
30 ml
Catalog # SPVAC00827P09028
Supplier: GRAVIS
Jump to:
Product Details & Documents
Glass, clear, amber or yellow.
- Neck Ø: 28 mm
- Good chemical and temperature resistance
- Amber coloured to protect light sensitive substances
Choice of various screw caps.
: Supplied without screw caps - please order separately.
: Packaged in impermeable plastic packs.
Specifications
- Capacity:30 ml
- Seq:11



