Orders LinkContactUsLinkComponent
Scissors, soft-cut
Scissors, soft-cut
213 mm
Catalog # WEDO9898
Supplier:   WEDO WERNER DORSCH
Scissors, soft-cut
Catalog # WEDO9898
Supplier:  WEDO WERNER DORSCH
Scissors, soft-cut
Scissors, soft-cut
213 mm
Price
22,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel with plastic handles. Universal scissors for a wide variety of uses.
  • Ergonomically shaped, rubberised grip areas for comfortable handling
  • Precisely ground blades for the perfect cut
  • With screw for readjusting
Specifications
  • Length:
    213 mm