Adhesive seals
EASYseal adhesive film, non sterile, 80×140 mm, Transparent, -
Catalog # GREI676001BE
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Plate ealer, for microplates, 120x80mm, coding, 200 pcs 1 * 200 items Catalog #GREI676004_200
Product GREI676004_200 is similar to GREI676001BE but manufacturer and specifications may vary.
Product Details & Documents
These adhesive seals offer an alternative to heat seals or sealing mats. EASYseal™ is a transparent adhesive film designed to cover microplates to protect samples from evaporation and contamination. It is suitable for routine laboratory use, including ELISA applications. BREATHseal™ is a gas permeable, pierceable membrane coated with an acrylic adhesive. It is ideal for bacterial, yeast and cell culture applications, assuring the protection of cell contents from contamination whilst maintaining oxygenation.
- Free from RNase, DNase and human DNA
- Non pyrogenic
Specifications
- Optical property:Transparent
- Sterile:-
- Description:EASYseal adhesive film, non sterile, 80×140 mm
- Seq:03