Orders LinkContactUsLinkComponent
Staining jars
Staining tray, pressed glass, - mm
Catalog # MARI4200005
Supplier:   MARIENFELD
Staining jars
Catalog # MARI4200005
Supplier:  MARIENFELD
Staining jars
Staining jars
Staining tray, pressed glass, - mm
Price
198,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Staining set consists of glass dish and staining jar made of soda lime glass, and staining tray made of either pressed glass or stainless steel.
These staining tray hold up to 10 slides (76×26 mm).
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.
Specifications
  • Description:
    Staining tray, pressed glass
  • W×D×H:
    - mm