Orders LinkContactUsLinkComponent
Height adjustable worktable
Height adjustable worktable
Concept motor table, (motorised height adjustment) ; control panel with three memory settings, 1500×750×700/1100 mm
Catalog # SOVEC10341037
Supplier:   TRESTON
Height adjustable worktable
Catalog # SOVEC10341037
Supplier:  TRESTON
Height adjustable worktable
Height adjustable worktable
Concept motor table, (motorised height adjustment) ; control panel with three memory settings, 1500×750×700/1100 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
These worktables are a suitable solution for industrial laboratories and research and development centers. They are designed to accommodate the necessary measurement equipment and other tools ergonomically at a safe distance for use. These laboratory benches fit also to education purposes. Modularity and accessories are individually adjustable according the task and human requirements.
  • Robust, elegant, and very flexible
  • Easy to adapt to individual needs, new equipment, and changes in the working environment
  • With durable table frame
  • Leveling glides
Ordering information: Table top sold separately; contact your Avantor representative for more information.
Specifications
  • Description:
    Concept motor table, (motorised height adjustment) ; control panel with three memory settings
  • W×D×H:
    1500×750×700/1100 mm