5l/min, >99.995%*, <0.1ppm hc, without compressor, 5.7 bar g (83 psi g) 1 * 1 items
PARK76-98UK
5l/min, >99.995%*, <0.1ppm hc, without compressor, 5.7 bar g (83 psi g) 1 * 1 items
PARK76-98UK
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection