Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Manual cell counters

Manual cell counter, 4
  MOIN1494MW
 :   MOINEAU
Manual cell counters
  MOIN1494MW
 :  MOINEAU
Manual cell counters
Manual cell counters

Manual cell counter, 4

 
402,00 €
   
1 * 1 ST
 

This manual cell counter can be used in combination of up to five counters.

  • Four digit display (4,3×3,5 mm) with reset knob
  • Use finger pressure
  • Can be fixed in position

W×D×H: 37,6×0,4×38,1 mm

 
  • Description:
    Manual cell counter
  • Seq:
    003
  • Array:
    4
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.