Stoppers, for test tubes
Green, 11 - 12 mm
Catalog # RATI351490G
Supplier: RATIOLAB
Aller à :
Product Details & Documents
PE.
- Very tight seal due to two sealing rings
- Available in natural and four more colors
Spécifications
- Ø bottom:11 - 12 mm
- Colour:Green
- Seq:03