Microplates, 96-well
96 Well plate, Clear, 96, +
RATI6018113
: RATIOLAB
Documents
- No. of wells:96
- Sterile:+
- Colour:Clear
- Description:96 Well plate
- Cat. No.:RATI6018113
- Model:Micro Test Plates, volume ∼300 μl
- Well profile:U-bottom