Orders LinkContactUsLinkComponent
Funnels, powder
100 mm, 35 mm, 15 mm
  KAVA632413102100
 :   Kavalier
Funnels, powder
  KAVA632413102100
 :  Kavalier
Funnels, powder
Funnels, powder
100 mm, 35 mm, 15 mm
44,30 €
1 * 1 ST
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
 : Ø = internal stem diameter.
  • Ø:
    15 mm
  • Funnel O-Ø:
    100 mm
  • Stem length:
    35 mm