Orders LinkContactUsLinkComponent
Sampler, StickProof
Sampler, 1100 mm, 25 mm, ≤50 ml
  300-0117
 :   BURKLE
Sampler, StickProof
  300-0117
 :  BURKLE
Sampler, StickProof
Sampler, StickProof
Sampler, 1100 mm, 25 mm, ≤50 ml
892,00 €
1 * 1 ST
Stainless steel sampler for powders. The conical inlet allows for collection of different sample sizes, up to approximately 50 ml. Constructed without edges, grooves or crevices for easy cleaning. The sample is collected directly into the sampling bag which is held in place with the included clamp. StickProof borehole diameter 25 mm; chamber length 120 mm. Sampling bags: 170×120 mm for 400 ml.
  • Especially slender tip can be inserted easily and deeply
  • Electropolished surface
  • Variable sample size up to 50 ml
  • Stainless steel V4A (1.4404)/AISI 316 L
 : Complete with clamp and 100 sampling bags PE.
  • Ø:
    25 mm
  • Volume:
    ≤50 ml
  • Description:
    Sampler
  • Length:
    1100 mm
Accessories
  •  
    Capacity
    Thickness
    Dimensions
  • 113-5415
    140 ml
    0,05
    80×120 mm
    1 * 100 ST - 13,20 €
  •  
    Description
  • 113-5414
    Packaging bags with closure (120×170 mm), 400 ml
    1 * 100 ST - 20,50 €
  •  
    Capacity
    Thickness
    Dimensions
  • 113-5413
    6500 ml
    0,05
    300×400 mm
    1 * 100 ST - 54,80 €
  •  
    Capacity
    Thickness
    Dimensions
  • 113-5412
    2500 ml
    0,05
    200×300 mm
    1 * 100 ST - 36,90 €
  •  
    Capacity
    Thickness
    Dimensions
  • 113-5411
    1000 ml
    0,05
    160×220 mm
    1 * 100 ST - 27,00 €
  •  
    Capacity
    Thickness
    Dimensions
  • BURK2348-0009
    4500 ml
    0,05
    250×350 mm
    1 * 100 ST - 47,80 €
  •  
    Capacity
    Thickness
    Dimensions
  • BURK2348-0002
    80 ml
    0,05
    70×100 mm
    1 * 100 ST - 11,10 €
  •  
    Capacity
    Thickness
    Dimensions
  • BURK2348-0007
    1500 ml
    0,05
    180×250 mm
    1 * 100 ST - 29,70 €
  •  
    Capacity
    Thickness
    Dimensions
  • BURK2348-0004
    300 ml
    0,05
    100×150 mm
    1 * 100 ST - 16,20 €
  •  
    Capacity
    Thickness
    Dimensions
  • BURK2348-0001
    100 ml
    0,05
    60×130 mm
    1 * 100 ST - 11,70 €