Dropping bottles, LDPE
1000 ml, 295 mm, 90 mm
Catalog # LAMA107
Supplier: LAMAPLAST
Aller à :
Product Details & Documents
LDPE, translucent, narrow mouth
- With cap and dosing tip
- Suitable for foodstuffs
- Good chemical and temperature resistance
Documents
TDS Drop Bottles, PL 101 - 107.pdf
Datasheet
Spécifications
- Ext.Ø:90 mm
- Capacity:1000 ml
- Height:295 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



