Orders LinkContactUsLinkComponent
Tubes and systems for active sampling
Tubes and systems for active sampling
Activated charcoal tubes, G
Catalog # DRAE6728831
Supplier:   Dräger
Tubes and systems for active sampling
Catalog # DRAE6728831
Supplier:  Dräger
Tubes and systems for active sampling
Tubes and systems for active sampling
Activated charcoal tubes, G
Price
77,60 €
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
  • High precision guaranteed
  • Very reliable in use
  • Improved colour depth and contrast using an optimised production method
Spécifications
  • Type:
    G
  • Description:
    Activated charcoal tubes
  • Seq:
    06