ContactUsLinkComponent
Single-channel RDO meter with InLab OptiOx (ISM, IP67 and 1.8 m fixed cable) complete with batteries, wrist strap, operating instr uctions, test certificate and declaration of conformity 1 * 1 items
Catalog # 664-0069
Supplier:  Mettler - Toledo
Single-channel RDO meter with InLab OptiOx (ISM, IP67 and 1.8 m fixed cable) complete with batteries, wrist strap, operating instr uctions, test certificate and declaration of conformity 1 * 1 items
Catalog # 664-0069
Supplier:  Mettler - Toledo