Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
96-well microplate, F-bottom (standard), MICROLON® 200, individually packed, 25 - 340 µl, 382 µl
Catalog # GREI655001-000
   Low Manufacturing Impact
Supplier:   Greiner Bio-One
Microplates, 96-well
Catalog # GREI655001-000
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Microplates, 96-well
Microplates, 96-well
96-well microplate, F-bottom (standard), MICROLON® 200, individually packed, 25 - 340 µl, 382 µl
Price
96,40 €
Log in for availability
1 * 40 ST
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
  • Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifications
  • Description:
    96-well microplate, F-bottom (standard), MICROLON® 200, individually packed
  • Seq:
    04
  • Working volume:
    25 - 340 µl
  • Total volume:
    382 µl
  • Cat. No.:
    GREI655001-000
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact