Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage tubes, barcoded, Corning®

Storage tubes, barcoded, Corning®

384× 2D barcoded storage tubes, round, without plug caps, 110 µl, Black, racked, do 90 µl
Catalogus # 479-0972
Leverancier:   Corning
Storage tubes, barcoded, Corning®
Catalogus # 479-0972
Leverancier:  Corning
Storage tubes, barcoded, Corning®
Storage tubes, barcoded, Corning®

384× 2D barcoded storage tubes, round, without plug caps, 110 µl, Black, racked, do 90 µl

Price
2 410,00 €
Log in voor beschikbaarheid
1 * 7.680 ST
Product Details & Documents

PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.

  • Maximum information, 14×14 dot 2D barcode
  • Barcode stability, laser-etched, not an attached label
  • Compound compatibility, polypropylene construction throughout, making it inert

Documents

Specificaties
  • Packed:
    racked
  • Recommended working volume:
    do 90 µl
  • Capacity:
    110 µl
  • Colour:
    Black
  • Description:
    384× 2D barcoded storage tubes, round, without plug caps
Accessories

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0248
    Empty racks with lids for 384 round or square storage tubes
    Blue
    -
    Log in voor beschikbaarheid
    1 * 20 ST - 219,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0247
    384× storage tube plug cap on mats for round tube, 384 caps per mat
    Orange
    384 caps per mat
    Log in voor beschikbaarheid
    1 * 38.400 ST - 4 260,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0246
    384× storage tube plug cap for round tube
    Orange
    bulk pack
    Log in voor beschikbaarheid
    1 * 38.400 ST - 3 040,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0245
    Empty racks with lids for 96 screw capped or Thermoplastic Elastomers (TPE) capped storage tubes
    Natural
    -
    Log in voor beschikbaarheid
    1 * 10 ST - 81,10 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0243
    Thermoplastic elastomer (TPE) 96 storage tube septum cap
    White
    bulk packed
    Log in voor beschikbaarheid
    1 * 9.600 ST - 590,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0244
    Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat
    White
    96 caps per mat
    Log in voor beschikbaarheid
    1 * 4.800 ST - 826,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0242
    96× storage tube screw cap, with O-Ring, on mat, clear with cover
    Orange
    96 caps per mat
    Log in voor beschikbaarheid
    1 * 4.800 ST - 3 400,00 €

Productinformatie

  • Catalogus #
    Description
    Colour
    Packed
    Beschikbaarheid
    Price
  • 731-0241
    96× screw caps for storage tubes 1,3 ml, with O-Ring
    Orange
    bulk packed
    Log in voor beschikbaarheid
    1 * 4.800 ST - 2 830,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.