MSD Read Buffer T (4x
Catalog # MESOR92TC-3
Supplier: MESO SCALE DISCOVERY
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
MicroWell™ Plates, 96 well, Nunc-Immuno™
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Mouse GFAP ELISA Kit
Supplier: Abcam
TOTAL PROTEIN UC STANDARD FS 6X3ML 1 * 1 KIT
Supplier: DIAGNOSTIC SYSTEMS DIASYS