WHEATON® Rubber Stoppers
Stopper, natural red rubber, 5,0 mm, 11,0 mm
Catalog # 215-4708
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Stoppers of various styles, sizes and rubber formulations.
- Manufactured from the highest quality raw materials
- Components manufactured to yield exceptional dimensional stability
- Autoclavable
Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish.
Snap-on style provides maximum sealing for vials with a blow-back glass finish.
Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure.
Specifications
- Ø bottom:5,0 mm
- Ø top:11,0 mm
- Description:Stopper, natural red rubber
- Seq:01
- Cat. No.:215-4708
- Durometer hardness:39
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



