Biotin kits, Micro HOOK™
HOOK™-sulfo-NHS-Biotin Kit, 1 KIT
Catalog # 786-694
Supplier: G-Biosciences
Jump to:
Product Details & Documents
The micro HOOK™ Biotin kits are designed to label small amounts of proteins, with each kit designed for 8 to 10 labellings of 50 to 250 µg protein/reaction.
- Micro kit for labelling protein primary amines
- Optimizer Buffer™ for improved coupling efficiency
- Gel filtration columns for rapid (<10 minute) purification
- Labels 50 to 250µg protein/reaction
- Suitable for 8 to 10 couplings
Kits contain the following Biotin reagents
HOOK™ Sulfo-NHS-Biotin - amine reactive reagent, shortest spacer arm
HOOK™ Sulfo-NHS-LC-Biotin - amine reactive reagent, longer spacer arm
HOOK™ Sulfo-NHS-SS-Biotin - cleavable, amine reactive reagent
HOOK™ NHS-dPEG₄-Biotin - amine reactive pegylated reagent
Documents
Specifications
Specifications
- Pk:1 KIT
- Description:HOOK™-sulfo-NHS-Biotin Kit