Pipette tips
Extended length (91 mm), 1 - 200 µl, -, Transparent, Pipetman, Eppendorf, Nichiryo, Oxford, Costar, Volac, Rainin, Finnpipette, Bulk
Catalog # DELT200026
Supplier: DELTALAB
Jump to:
Product Details & Documents
Documents
Specifications
- Packed:Bulk
- Sterile:-
- Volume:1 - 200 µl
- Seq:06
- Tip style:Extended length (91 mm)
- Tip colour:Transparent
- For use with:Pipetman, Eppendorf, Nichiryo, Oxford, Costar, Volac, Rainin, Finnpipette