Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 9,180, 250 mL
N° de catalogue METT51350056
Fournisseur:   Mettler - Toledo
pH buffer solutions, NIST/DIN
N° de catalogue METT51350056
Fournisseur:  Mettler - Toledo
Numéro CAS:  
pH buffer solutions, NIST/DIN

NIST/DIN buffer solution, 9,180, 250 mL

Numéro CAS:  
Prix
30,50 €
Log in for availability
1 * 250 mL
Product Details & Documents

The NIST/DIN pH buffers are specified to three decimal places, e.g. 9.180, offering the highest possible pH precision.

A detailed test certificate is available for every bottle and guarantees the tested values and traceability.

 : NIST traceable.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    NIST/DIN buffer solution
  • pH value:
    9,180

Specifications