Order Entry
Belgium
Orders LinkContactUsLinkComponent
Redox buffer solutions

Redox buffer solutions

Redox buffer, 220 mV, 7, 6×250 ml, 6
Catalog # METT51350062
Supplier:   Mettler - Toledo
Redox buffer solutions
Catalog # METT51350062
Supplier:  Mettler - Toledo
CAS Number:  
Redox buffer solutions

Redox buffer, 220 mV, 7, 6×250 ml, 6

CAS Number:  
Price
178,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

Redox buffer solutions are used for verification of all common redox sensors.

  • Temperature dependent
  • Used in quality assurance systems

A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.

Delivery information: Supplied with a quality inspection certificate.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pk:
    6
  • Description:
    Redox buffer, 220 mV
  • Pk Info:
    6×250 ml
  • pH value:
    7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.