Order Entry
Belgium
Orders LinkContactUsLinkComponent
Maintenance solutions for electrodes

Maintenance solutions for electrodes

Cleaning solution thiourea, 250 ml, 250 mL
Catalog # METT51350102
Supplier:   Mettler - Toledo
Maintenance solutions for electrodes
Catalog # METT51350102
Supplier:  Mettler - Toledo
CAS Number:  
Maintenance solutions for electrodes

Cleaning solution thiourea, 250 ml, 250 mL

CAS Number:  
Price
30,70 €
Log in for availability
1 * 250 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Cleaning solution thiourea, 250 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.