pH buffer solutions
Technical buffer solution, 250 ml, 11,00, 250 mL
:
- Pk:250 mL
- Description:Technical buffer solution, 250 ml
- pH value:11,00
Magnesium sulphate heptahydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
#: 25165.292
: VWR Chemicals
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 21333.296
: VWR Chemicals
Potassium dihydrogen phosphate 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
#: 26936.293
: VWR Chemicals
Glass microfibre filters without binder, grade GF/C, Whatman™
#: 513-5227
: Whatman products (Cytiva)
Water, HiPerSolv CHROMANORM®, super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
#: 83650.320
: VWR Chemicals
Hydrochloric acid 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., NF, Reag. USP volumetric solution
#: 30024.290
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
#: 20066.296
: VWR Chemicals



