Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions

Technical buffer solution, 6×250 ml, 4,01, 6
Catalog # METT51350018
Supplier:   Mettler - Toledo
pH buffer solutions
Catalog # METT51350018
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 6×250 ml, 4,01, 6

CAS Number:  
Price
91,10 €
Log in for availability
1 * 6 ST
Specifications

Specifications

  • Pk:
    6
  • Description:
    Technical buffer solution, 6×250 ml
  • pH value:
    4,01

Specifications