Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, certified (DKD)

pH buffer solutions, certified (DKD)

DKD certified buffer solution, 9,21, 1× 250 ml, 250 mL
N° de catalogue METT51350036
Fournisseur:   Mettler - Toledo
pH buffer solutions, certified (DKD)
N° de catalogue METT51350036
Fournisseur:  Mettler - Toledo
Numéro CAS:  
pH buffer solutions, certified (DKD)

DKD certified buffer solution, 9,21, 1× 250 ml, 250 mL

Numéro CAS:  
Prix
27,80 €
Log in for availability
1 * 250 mL
Product Details & Documents

 : The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the ideally suitable products for regulated industries. DKD is accepted by other accreditation bodies within the scope of the European Cooperation for Accreditation (EA) and the International Laboratory Accreditation Cooperation (ILAC).

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    DKD certified buffer solution
  • Pk Info:
    1× 250 ml
  • pH value:
    9,21

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.