Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, certified (DKD)

pH buffer solutions, certified (DKD)

DKD certified buffer solution, 9,21, 6× 250 ml, 6
Catalog # METT51350046
Supplier:   Mettler - Toledo
pH buffer solutions, certified (DKD)
Catalog # METT51350046
Supplier:  Mettler - Toledo
CAS Number:  
pH buffer solutions, certified (DKD)

DKD certified buffer solution, 9,21, 6× 250 ml, 6

CAS Number:  
Price
101,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

Certifications: The DKD (Deutscher Kalibrierdienst, German Calibration Service) certified buffers are the ideally suitable products for regulated industries. DKD is accepted by other accreditation bodies within the scope of the European Cooperation for Accreditation (EA) and the International Laboratory Accreditation Cooperation (ILAC).

Specifications

Specifications

  • Pk:
    6
  • Description:
    DKD certified buffer solution
  • Pk Info:
    6× 250 ml
  • pH value:
    9,21

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.