Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 6×250 ml, 9,21, Colourless, 250 ml, 6
Catalogus # METT51350022
Leverancier:   Mettler - Toledo
pH buffer solutions
Catalogus # METT51350022
Leverancier:  Mettler - Toledo
CAS-nummer:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 6×250 ml, 9,21, Colourless, 250 ml, 6

CAS-nummer:  
Price
91,10 €
Log in voor beschikbaarheid
1 * 6 ST
Product Details & Documents
Specificaties

Specificaties

  • Pk:
    6
  • Colour:
    Colourless
  • Description:
    Technical buffer solution, 6×250 ml
  • Pk Info:
    250 ml
  • pH value:
    9,21

Specificaties