Orders LinkContactUsLinkComponent
Microplate Lids
Microplate lid for MTP/DWP, sterile, (5 bags×16), Sterile
Catalog # 737-0199
   Low Manufacturing Impact
Supplier:   EPPENDORF
Microplate Lids
Catalog # 737-0199
Supplier:  EPPENDORF
   Low Manufacturing Impact
Microplate Lids
Microplate Lids
Microplate lid for MTP/DWP, sterile, (5 bags×16), Sterile
Price
174,00 €
Log in for availability
1 * 80 ST
Product Details & Documents
Protect your samples with Eppendorf® Microplate Lids, perfect for incubation and short-term storage. Stable, flexible protection available in 'PCR Clean' and 'Sterile' grades, ensuring your samples stay pure and uncontaminated.
  • Stable, flexible protection of samples during incubation and short-term storage
Specifications
  • Sterility:
    Sterile
  • Description:
    Microplate lid for MTP/DWP, sterile, (5 bags×16)
  • Seq:
    02
  • Cat. No.:
    737-0199
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact