Orders LinkContactUsLinkComponent
Screw caps
GL 18
  VITL102597
 :   VITLAB
Screw caps
  VITL102597
 :  VITLAB
Screw caps
Screw caps
GL 18
8,10 €
1 * 1 ST
PFA, transparent
  • Especially suitable for use in trace analysis
  • Ideal for sensitive and valuable samples
  • High thermal stability, from –200 °C to +260 °C, autoclavable
  • Chemical inertness against nearly all chemicals
  • Easy to clean
For sealing all PFA containers with GL threads or buttress threads.
  • Thread:
    GL 18