Gel permeation chromatography columns, PolyPore™
Analytical, 5 µm, 7,50 mm, 300 mm
Catalog # VARIPL1113-6500
Supplier: VARIAN
Jump to:
Product Details & Documents
PolyPore columns results unrivalled resolution for the analysis of polymers with broad molecular weight distributions.
- Routine polymer analysis with very high resolution
- Wide operating range simplifies column choice
- Low particle size extracts maximum information from the analyte
Specifications
- Column Format:Analytical
- Ø int.:7,50 mm
- Particle Size:5 µm
- Length:300 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



