Order Entry
Belgium
Orders LinkContactUsLinkComponent
High efficiency GC columns

High efficiency GC columns

VF-1ms, 0,60 µm, 0,15 mm, 25 m
Catalog # VARICP8931
Supplier:   VARIAN
High efficiency GC columns
Catalog # VARICP8931
Supplier:  VARIAN
High efficiency GC columns
High efficiency GC columns

VF-1ms, 0,60 µm, 0,15 mm, 25 m

Price
877,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

High efficiency 0,15 and 0,18 mm Int.Ø columns are a simple and cost effective way to increase sample throughput with existing GC or GC/MS instruments.

  • Increase sample throughput and productivity while maintaining resolution
  • Reduce carrier gas usage and cost per analysis
  • Operates with He or H₂ carrier gas
  • Available in more than 20 stationary phases

This leading-edge column technology is ideal for applications that require faster run times, such as high-throughput screening, fast process monitoring, and fast method development. Agilent High Efficiency GC columns can reduce sample run time by 50% or more without compromising resolution.

Specifications
  • Film thickness:
    0,60 µm
  • Ø int.:
    0,15 mm
  • Length:
    25 m
  • Phase:
    VF-1ms
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.