Torches
Radial torches, high solids torch with patented injector design, 1,8 mm ID
Catalog # VARI2010117500
Supplier: VARIAN
Jump to:
Product Details & Documents
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.
Specifications
- Description:Radial torches, high solids torch with patented injector design, 1,8 mm ID
- Seq:09
Accessories
Product Information
- Catalog #AvailabilityPrice
- Spare quartz torch bonnet for radial ICP-OESSpare quartz torch bonnet for radial ICP-OESVARI2010070790Log in for availability1 * 1 ST - 161,00 €
Product Information
- Catalog #AvailabilityPrice
- Torch alignment tool for radial ICP-OES
Product Information
- Catalog #AvailabilityPrice
- GazFit fittings for one-piece and semi-demountable ICP-OES torchesGazFit fittings for one-piece and semi-demountable ICP-OES torchesVARI7910051290Log in for availability1 * 1 ST - 221,00 €
Product Information
- Catalog #AvailabilityPrice
- Transfer tube FEP for axial ICP
Product Information
- Catalog #AvailabilityPrice
- Transfer tube FEP Sturman-Masters spray chamber for radial ICPTransfer tube FEP Sturman-Masters spray chamber for radial ICPVARI7910051490Log in for availability1 * 1 ST - 221,00 €
Product Information
- Catalog #AvailabilityPrice
- Kit Torch Fittings
Product Information
- Catalog #AvailabilityPrice
- Pre-optics cone for Liberty Series axial ICP-OESPre-optics cone for Liberty Series axial ICP-OESVARI3610007300Log in for availability1 * 1 ST - 818,00 €
Product Information
- Catalog #AvailabilityPrice
- Esteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OESEsteele cleaning powder, for cleaning interface cone on axially-viewed ICP-OESVARI8110002800Log in for availability1 * 1 ST - 52,00 €
Product Information
- Catalog #AvailabilityPrice
- Spare semi-demountable quartz torch body for radial ICP-OESSpare semi-demountable quartz torch body for radial ICP-OESVARI2010072900Log in for availability1 * 1 ST - 1 060,00 €
Product Information
- Catalog #AvailabilityPrice
- Quartz injector tube, 0,8 mm
Product Information
- Catalog #AvailabilityPrice
- Spare semi-demountable quartz torch body for axial ICP-OESSpare semi-demountable quartz torch body for axial ICP-OESVARI2010083400Log in for availability1 * 1 ST - 1 060,00 €
Product Information
- Catalog #AvailabilityPrice
- Quartz injector tube, 1,4 mm
Product Information
- Catalog #AvailabilityPrice
- Injector holder kit, includes O-ring
Product Information
- Catalog #AvailabilityPrice
- O-rings kit