Blocking buffer, fish gelatin
Fish gelatin blocking buffer, 10% solution, 100 mL
Catalog # 22010.
Supplier: Biotium
Aller à :
Product Details & Documents
Fish gelatin is commonly used in blocking solutions to minimise background noise in immunodetection procedures such as Western Blotting and immunostaining. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.
Spécifications
Spécifications
- Pk:100 mL
- Description:Fish gelatin blocking buffer, 10% solution
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



